Learn More
Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | 99.8% | C9H17Cl2N5O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA0124A10MM/1ML
Encompass_Preferred
Sapropterin dihydrochloride is an orally active phenylalanine hydroxylase (PAH) cofactor. It is effective in reducing blood phenylalanine concentrations and can be used in the study of phenylketonuria (PKU). This compound enhances residual phenylalanine hydroxylase activity to increase phenylalanine oxidation and can also aggravate experimental autoimmune encephalomyelitis (EAE). For research use only.
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Used in the study of phenylketonuria (PKU)
  • Enhances residual phenylalanine hydroxylase activity
Catalog No. 50-004-32349