Learn More
Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | 99.8% | C9H17Cl2N5O3 | 25 MG
SDP
Catalog No. 5000358894
Encompass_Preferred
Sapropterin dihydrochloride is an orally active phenylalanine hydroxylase (PAH) cofactor. It is effective in reducing blood phenylalanine concentrations and can be used in the study of phenylketonuria (PKU). Additionally, it has been shown to aggravate experimental autoimmune encephalomyelitis (EAE).
  • Enhances residual phenylalanine hydroxylase activity to increase phenylalanine oxidation.
  • Aggravates experimental autoimmune encephalomyelitis (EAE) in SJL/J mice.
  • Increases clinical EAE scores and elevates plasma long-chain ceramide levels.
  • Reduces linolenic acid levels and increases spinal cord T-cell infiltration.
Catalog No. 50-003-58894 Supplier Medchemexpress LLC Supplier No. 25MGHYA0124AR
Sapropterin dihydrochloride is an orally active phenylalanine hydroxylase (PAH) cofactor. It is effective in reducing blood phenylalanine concentrations and can be used in the study of phenylketonuria (PKU). Additionally, it has been shown to aggravate experimental autoimmune encephalomyelitis (EAE).
  • Enhances residual phenylalanine hydroxylase activity to increase phenylalanine oxidation.
  • Aggravates experimental autoimmune encephalomyelitis (EAE) in SJL/J mice.
  • Increases clinical EAE scores and elevates plasma long-chain ceramide levels.
  • Reduces linolenic acid levels and increases spinal cord T-cell infiltration.