Learn More
Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | 99.96% | C9H17Cl2N5O3 | 50 MG
SDP
Catalog No. 5000432474
Encompass_Preferred
Sapropterin dihydrochloride is an analytical standard used for research and analytical applications. It functions as an orally active phenylalanine hydroxylase (PAH) cofactor, helping to reduce blood phenylalanine concentrations. This compound is suitable for studying phenylketonuria (PKU). It is an analytical standard grade, commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Useful in the study of phenylketonuria (PKU)
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-32474 Supplier Medchemexpress LLC Supplier No. HYA0124AR50MG
Sapropterin dihydrochloride is an analytical standard used for research and analytical applications. It functions as an orally active phenylalanine hydroxylase (PAH) cofactor, helping to reduce blood phenylalanine concentrations. This compound is suitable for studying phenylketonuria (PKU). It is an analytical standard grade, commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Useful in the study of phenylketonuria (PKU)
  • Suitable for HPLC, GC, and MS applications