Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sapropterin dihydrochloride | 69056-38-8 | MFCD00891665 | 314.17 | C9H17Cl2N5O3 | 10 MG
SDP

Catalog No. 5000299282
Encompass_Preferred

Sapropterin dihydrochloride is the dihydrochloride salt of sapropterin ((6R)-BH4), supplied for research use as a PAH cofactor and analytical standard for studies of phenylketonuria and phenylalanine metabolism. It is provided as a solid and is intended for in vitro and analytical applications.

  • Dihydrochloride salt form for improved handling and stability.
  • Used as a cofactor in phenylalanine hydroxylase studies.
  • Suitable for phenylketonuria and metabolic pathway research.
  • Molecular formula C9H17Cl2N5O3; molecular weight 314.17 g/mol.
  • Available in small research pack sizes suitable for assay and analytical work.

Catalog No. 50-002-99282 Supplier Medchemexpress LLC Supplier No. HYA0124AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sapropterin dihydrochloride is the dihydrochloride salt of sapropterin ((6R)-BH4), supplied for research use as a PAH cofactor and analytical standard for studies of phenylketonuria and phenylalanine metabolism. It is provided as a solid and is intended for in vitro and analytical applications.

  • Dihydrochloride salt form for improved handling and stability.
  • Used as a cofactor in phenylalanine hydroxylase studies.
  • Suitable for phenylketonuria and metabolic pathway research.
  • Molecular formula C9H17Cl2N5O3; molecular weight 314.17 g/mol.
  • Available in small research pack sizes suitable for assay and analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.