Learn More
Medchemexpress LLC Sapropterin dihydrochloride (standard) | 69056-38-8 | 100.0% | C9H17Cl2N5O3 | 5 MG
SDP
Catalog No. 5000433389
Encompass_Preferred
Sapropterin dihydrochloride (Standard) is the analytical standard of Sapropterin (dihydrochloride), intended for research and analytical applications. It functions as an orally active phenylalanine hydroxylase (PAH) cofactor, reducing blood phenylalanine concentrations, and is used in the study of phenylketonuria (PKU). Additionally, it can aggravate experimental autoimmune encephalomyelitis (EAE).
  • Analytical standard for research and analytical applications
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Used in the study of phenylketonuria (PKU)
  • Can aggravate experimental autoimmune encephalomyelitis (EAE)
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-33389 Supplier Medchemexpress LLC Supplier No. HYA0124AR5MG
Sapropterin dihydrochloride (Standard) is the analytical standard of Sapropterin (dihydrochloride), intended for research and analytical applications. It functions as an orally active phenylalanine hydroxylase (PAH) cofactor, reducing blood phenylalanine concentrations, and is used in the study of phenylketonuria (PKU). Additionally, it can aggravate experimental autoimmune encephalomyelitis (EAE).
  • Analytical standard for research and analytical applications
  • Orally active phenylalanine hydroxylase (PAH) cofactor
  • Reduces blood phenylalanine concentrations
  • Used in the study of phenylketonuria (PKU)
  • Can aggravate experimental autoimmune encephalomyelitis (EAE)
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS