Learn More
AdipoGen Saquayamycin B1
SDP
Catalog No. 502561961
Encompass_Preferred
Chemical. CAS 99260-68-1. Formula C31H32O12. MW 596.6. Isolated from Streptomyces nodosus. Angucycline antibiotic. Antibacterial. Antitumor compound. Active against Gram-positive bacteria and P388 leukaemia cells. Farnesyl-protein transferase inhibitor. Inhibitor of nitric oxide synthase NOS shown for Saquayamycin A1.
Catalog No. 50-256-1961 Supplier AdipoGen Supplier No. BVT0382C500
Chemical. CAS 99260-68-1. Formula C31H32O12. MW 596.6. Isolated from Streptomyces nodosus. Angucycline antibiotic. Antibacterial. Antitumor compound. Active against Gram-positive bacteria and P388 leukaemia cells. Farnesyl-protein transferase inhibitor. Inhibitor of nitric oxide synthase NOS shown for Saquayamycin A1.