Learn More
Medchemexpress LLC Saquinavir mesylate | 149845-06-7 | 99.8% | 766.95 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17003100MG
Encompass_Preferred
Saquinavir mesylate is an orally active HIV protease inhibitor. It is used in AIDS research and exhibits anti-inflammatory activity, capable of inducing apoptosis in human red blood cells.
  • Orally active HIV protease inhibitor
  • Used in AIDS research
  • Possesses anti-inflammatory activity
  • Induces apoptosis of human red blood cells
  • Exhibits anti-HIV activity in mouse models
  • Protective effect in mouse models of lung injury
Catalog No. 50-003-91258