Learn More
Medchemexpress LLC Saquinavir mesylate | 149845-06-7 | 99.8% | 766.95 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY170035MG
Encompass_Preferred
Saquinavir mesylate (Ro 31-8959) is an orally active HIV protease inhibitor used in AIDS research. It also demonstrates anti-inflammatory activity and induces apoptosis of human red blood cells.
  • Orally active HIV protease inhibitor
  • Used in AIDS research
  • Has anti-inflammatory activity
  • Induces apoptosis of human red blood cells
  • Exhibits anti-HIV activity in the thy/liv-SCID-hu mouse model
  • Has a protective effect in the mouse model of lung injury induced by hepatic ischemia-reperfusion (I/R)
Catalog No. 50-003-90684