Learn More
Apexbio Technology LLC Saquinavir mesylate 149845-06-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064120
Encompass_Preferred
Saquinavir mesylate is a reversible inhibitor of HIV protease a retroviral aspartic protease required for viral maturation and replication It targets the active site of HIV-1 and HIV-2 proteases preventing enzymatic cleavage of polyprotein precursors thereby suppressing formation of mature viral particles Saquinavir inhibits HIV-1 protease with a reported Ki value of approximately 0 12 nM In cultured human neuronal cells it also reduces -amyloid (A 40) levels and inhibits -secretase (BACE1) activity at 50 M concentration it reduces neuronal A production by approximately 90% Saquinavir is commonly employed in HIV/AIDS research and has potential in Alzheimer s disease models for studying the modulation of amyloid peptide synthesis pathways
Catalog No. 50-000-64120 Supplier Apexbio Technology LLC Supplier No. A37915.1
Saquinavir mesylate is a reversible inhibitor of HIV protease a retroviral aspartic protease required for viral maturation and replication It targets the active site of HIV-1 and HIV-2 proteases preventing enzymatic cleavage of polyprotein precursors thereby suppressing formation of mature viral particles Saquinavir inhibits HIV-1 protease with a reported Ki value of approximately 0 12 nM In cultured human neuronal cells it also reduces -amyloid (A 40) levels and inhibits -secretase (BACE1) activity at 50 M concentration it reduces neuronal A production by approximately 90% Saquinavir is commonly employed in HIV/AIDS research and has potential in Alzheimer s disease models for studying the modulation of amyloid peptide synthesis pathways