Learn More
Apexbio Technology LLC Saquinavir mesylate, 25mg. Cas: 149845-06-7 MFCD: MFCD00944907. HIV Protease Inhibitor
SDP
Supplier:  Apexbio Technology LLC A379125
Encompass_Preferred
Saquinavir is a potent inhibitor HIV protease with Ki value of 0.12 and < 0.1 nM for HIV-1 and HIV-2 protease[1].HIV protease is essential for release of mature viral and viral replication which is a retroviral aspartyl protease[2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4840