Learn More
Cayman Chemical SaquInavIrmesylate 10mg
SDP
Catalog No. 502893356
Encompass_Preferred
An HIV protease inhibitor (Kis = 0.12 and 50 = 2 nM); inhibits SARS-CoV-2 Mpro activity (IC50 = 9.92 µM)
Catalog No. 50-289-3356 Supplier Cayman Chemical Supplier No. 900189310 MG
An HIV protease inhibitor (Kis = 0.12 and 50 = 2 nM); inhibits SARS-CoV-2 Mpro activity (IC50 = 9.92 µM)