Learn More
Selleck Chemical LLC SAR-020106
SDP
Catalog No. 5000002882
Encompass_Preferred
SAR-020106 is an ATP-competitive potent and selective CHK1 inhibitor with an IC50 of 13 3 nM
Catalog No. 50-000-02882 Supplier Selleck Chemical LLC Supplier No. S77405MG
SAR-020106 is an ATP-competitive potent and selective CHK1 inhibitor with an IC50 of 13 3 nM