Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLPBIO TECHNOLOGY INC SAR-100842 10MM IN 1ML DMSO

Supplier:  GLPBIO TECHNOLOGY INC GC3178510MM

Encompass_Preferred

SAR-100842 10mM (in 1mL DMSO)

Catalog No. NC2307978


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.