Learn More
Medchemexpress LLC HY-100895 5mg Medchemexpress, SAR-20347 CAS:1450881-55-6 Purity:>98%
SDP
Catalog No. 501931991
Encompass_Preferred
Medchemexpress, HY-100895 5mg SAR-20347 CAS:1450881-55-6 SAR-20347 is an inhibitor of TYK2 , JAK1 , JAK2 and JAK3 with IC 50 s of 0.6, 23, 26 and 41 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1991 Supplier Medchemexpress LLC Supplier No. HY1008955MG
Medchemexpress, HY-100895 5mg SAR-20347 CAS:1450881-55-6 SAR-20347 is an inhibitor of TYK2 , JAK1 , JAK2 and JAK3 with IC 50 s of 0.6, 23, 26 and 41 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.