Learn More
Selleck Chemical LLC SAR-20347
SDP
Catalog No. NC2230825
Encompass_Preferred
Catalog No.S0437SAR-20347SIZE 5MGSAR-20347 is a potent inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50 of 0.6 nM, 23 nM, 26 nM and 41 nM, respectively. SAR-20347 inhibits TYK2- and JAK1-mediated IL-12 and IFN-? signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
This item is not returnable. View return policy
Catalog No. NC2230825 Supplier Selleck Chemical LLC Supplier No. S04375MG
Catalog No.S0437SAR-20347SIZE 5MGSAR-20347 is a potent inhibitor of TYK2, JAK1, JAK2 and JAK3 with IC50 of 0.6 nM, 23 nM, 26 nM and 41 nM, respectively. SAR-20347 inhibits TYK2- and JAK1-mediated IL-12 and IFN-? signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.