Learn More
TARGETMOL CHEMICALS INC SAR-260301 5MG
SDP
Catalog No. 502577324
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. SAR-260301 is an orally available and selective inhibitor of PI3Kβ (IC50: 23 nM). purity: 98%
Catalog No. 50-257-7324 Supplier TARGETMOL CHEMICALS INC Supplier No. T168415MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. SAR-260301 is an orally available and selective inhibitor of PI3Kβ (IC50: 23 nM). purity: 98%