Learn More
Medchemexpress LLC Sar-Arg-Val-Tyr-Val-His-NH2 | 2410957-04-7 | 96.6% | 742.87 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP238110MG
Encompass_Preferred
Sar-Arg-Val-Tyr-Val-His-NH2 is a Gq-biased agonist that exhibits 10-fold larger molecular efficacies at the AT1R-Gq fusion protein compared with the AT1R-βarr2 fusion protein. This compound is targeted at the Angiotensin Receptor and acts via the GPCR/G Protein pathway.
  • Gq-biased agonist
  • Exhibits 10-fold larger molecular efficacies at AT1R-Gq fusion protein
  • Targets Angiotensin Receptor
  • Acts via GPCR/G Protein pathway
Catalog No. 50-004-22474