Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sar-[D-Phe8]-des-Arg9-bradykinin | 126959-88-4 | MFCD05662308 | 100.0% | 975.10 | C47H66N12O11 | 25 MG
SDP

Catalog No. 5000244662
Encompass_Preferred

Sar-[D-Phe8]-des-Arg9-bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used in pharmacological research to study receptor activation and contractile responses. It is supplied as a white to off-white solid with high purity and a defined amino acid sequence.

  • High purity suitable for analytical and biological assays.
  • Molecular weight 975.10 and formula C47H66N12O11.
  • Sequence: {Sar}-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe.
  • Stable as powder when stored sealed at -80°C (2 years) or -20°C (1 year).
  • Available in multiple sizes, including 25 MG.

Catalog No. 50-002-44662 Supplier Medchemexpress LLC Supplier No. HY10329125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sar-[D-Phe8]-des-Arg9-bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used in pharmacological research to study receptor activation and contractile responses. It is supplied as a white to off-white solid with high purity and a defined amino acid sequence.

  • High purity suitable for analytical and biological assays.
  • Molecular weight 975.10 and formula C47H66N12O11.
  • Sequence: {Sar}-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe.
  • Stable as powder when stored sealed at -80°C (2 years) or -20°C (1 year).
  • Available in multiple sizes, including 25 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.