Learn More
Medchemexpress LLC Sar-[D-Phe8]-des-Arg9-bradykinin | 126959-88-4 | MFCD05662308 | 100.0% | 975.10 | C47H66N12O11 | 5 MG
SDP
Catalog No. 5000244663
Encompass_Preferred
Sar-[D-Phe8]-des-Arg9-bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used as a pharmacological tool in vascular and smooth muscle research. It is supplied as a lyophilized solid with high purity for in vitro bioassays.
  • Selective bradykinin B1 receptor agonist used in receptor pharmacology.
  • Lyophilized white solid suitable for in vitro studies.
  • High purity (≈99.96%) for reproducible assay results.
  • Resistant to aminopeptidase cleavage, increasing stability in biological assays.
  • Supplied as a 5 mg vial for assay development and small-scale experiments.
Catalog No. 50-002-44663 Supplier Medchemexpress LLC Supplier No. HY1032915MG
Sar-[D-Phe8]-des-Arg9-bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used as a pharmacological tool in vascular and smooth muscle research. It is supplied as a lyophilized solid with high purity for in vitro bioassays.
  • Selective bradykinin B1 receptor agonist used in receptor pharmacology.
  • Lyophilized white solid suitable for in vitro studies.
  • High purity (≈99.96%) for reproducible assay results.
  • Resistant to aminopeptidase cleavage, increasing stability in biological assays.
  • Supplied as a 5 mg vial for assay development and small-scale experiments.