Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sar-[D-Phe8]-des-Arg9-Bradykinin | 126959-88-4 | MFCD05662308 | 100.0% | 975.10 g·mol⁻¹ | C47H66N12O11 | 10 MG
SDP

Catalog No. 5000244666
Encompass_Preferred

Sar-[D-Phe8]-des-Arg9-Bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used in pharmacology research to study B1-mediated vascular contractility and inflammatory signaling. It is supplied as a white to off-white solid with defined sequence and characterized physicochemical properties for in vitro applications.

  • High purity (99.96%).
  • White to off-white solid appearance.
  • Molecular weight 975.10 g·mol⁻¹.
  • Sequence: {Sar}-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe.
  • CAS number 126959-88-4.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic.
  • Store sealed at -80°C (up to 2 years) or -20°C (up to 1 year).
  • Available in multiple mg sizes (5 mg-100 mg).

Catalog No. 50-002-44666 Supplier Medchemexpress LLC Supplier No. HY10329110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sar-[D-Phe8]-des-Arg9-Bradykinin is a synthetic peptide agonist of the bradykinin B1 receptor used in pharmacology research to study B1-mediated vascular contractility and inflammatory signaling. It is supplied as a white to off-white solid with defined sequence and characterized physicochemical properties for in vitro applications.

  • High purity (99.96%).
  • White to off-white solid appearance.
  • Molecular weight 975.10 g·mol⁻¹.
  • Sequence: {Sar}-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe.
  • CAS number 126959-88-4.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic.
  • Store sealed at -80°C (up to 2 years) or -20°C (up to 1 year).
  • Available in multiple mg sizes (5 mg-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.