Learn More
Aviva Systems Biology Corp SAR1B Antibody - middle region (ARP56243_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56243_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAR1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SAR1B. Alternate names: CMRD, GTBPB, SARA2, ANDDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61779