Learn More
Apexbio Technology LLC SAR245409 (XL765) 1349796-36-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061839
Encompass_Preferred
SAR245409 (XL765 CAS 1349796-36-6) is a selective dual inhibitor of phosphatidylinositol-3-kinase (PI3K) and mammalian target of rapamycin (mTOR) with high potency against PI3K (IC50 9 nM) It suppresses the PI3K/Akt/mTOR signaling cascade by preventing formation of phosphatidylinositol-3 4 5-triphosphate (PIP3) at the cell membrane and downstream phosphorylation of AKT p70S6 kinase and S6 SAR245409 demonstrates antitumor activity in cancer models exhibiting genetic alterations in PI3K pathways enhancing growth suppression and apoptosis It is commonly employed in preclinical studies evaluating PI3K/mTOR-targeted approaches in cancer therapy
Catalog No. 50-000-61839 Supplier Apexbio Technology LLC Supplier No. A56345.1
SAR245409 (XL765 CAS 1349796-36-6) is a selective dual inhibitor of phosphatidylinositol-3-kinase (PI3K) and mammalian target of rapamycin (mTOR) with high potency against PI3K (IC50 9 nM) It suppresses the PI3K/Akt/mTOR signaling cascade by preventing formation of phosphatidylinositol-3 4 5-triphosphate (PIP3) at the cell membrane and downstream phosphorylation of AKT p70S6 kinase and S6 SAR245409 demonstrates antitumor activity in cancer models exhibiting genetic alterations in PI3K pathways enhancing growth suppression and apoptosis It is commonly employed in preclinical studies evaluating PI3K/mTOR-targeted approaches in cancer therapy