Learn More
Apexbio Technology LLC SAR245409 (XL765), 25mg. Cas: 1349796-36-6 MFCD: MFCD18252649. mTOR/PI3K inhibitor
SDP
Supplier:  Apexbio Technology LLC A563425
Encompass_Preferred
SAR245409 (XL765) is a selective dual inhibitor of PI3K and mTOR (IC50= 9 nM for PI3K). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4958