Learn More
Selleck Chemical LLC SAR405 5mg 1523406-39-4
SDP
Supplier:  Selleck Chemical LLC S76825MG
Encompass_Preferred
SAR405 is a low-molecular-mass kinase inhibitor of PIK3C3/Vps34 (KD 1.5 nM) showing high selectivity and not be active up to 10 ?M on class I and class II PI3Ks as well as on mTOR. SAR405 prevents autophagy and synergizes with MTOR (mechanistic target of rapamycin) inhibition in tumor cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5122