Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SAR405 R enantiomer | 1946010-79-2 | 99.9% | 443.85 | C19H21ClF3N5O2 | 10MG
SDP

Catalog No. 5000235179
Encompass_Preferred

SAR405 R enantiomer is the less active stereoisomer of the PIK3C3/Vps34 inhibitor SAR405, provided as a solid for in vitro research use only. It is suited for comparative pharmacology and control studies and includes handling and storage guidance for reliable experimental use.

  • Less active enantiomer used as a comparative control in assays.
  • High purity (≈99.9%).
  • Solid, white to off-white appearance.
  • Molecular formula C19H21ClF3N5O2; molecular weight 443.85.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.
  • Intended for laboratory research use only.

Catalog No. 50-002-35179 Supplier Medchemexpress LLC Supplier No. HY12481A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SAR405 R enantiomer is the less active stereoisomer of the PIK3C3/Vps34 inhibitor SAR405, provided as a solid for in vitro research use only. It is suited for comparative pharmacology and control studies and includes handling and storage guidance for reliable experimental use.

  • Less active enantiomer used as a comparative control in assays.
  • High purity (≈99.9%).
  • Solid, white to off-white appearance.
  • Molecular formula C19H21ClF3N5O2; molecular weight 443.85.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Store powder at -20°C for long-term stability; store solutions at -80°C.
  • Intended for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.