Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SAR405 R enantiomer | 1946010-79-2 | 99.9% | 443.85 | C19H21ClF3N5O2 | 10MM 1ML
SDP

Catalog No. 5000239238
Encompass_Preferred

SAR405 R enantiomer is the less active enantiomer of SAR405, a selective inhibitor of PIK3C3/Vps34, supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO or as a solid, and is used as a reference or control in studies of autophagy, vesicle trafficking, and related signaling pathways.

  • Less active enantiomer of a PIK3C3/Vps34 inhibitor.
  • High purity (~99.9%) for reliable experimental controls.
  • Available as 10 mM solution in DMSO and multiple solid package sizes.
  • Soluble in DMSO at 100 mg/mL; ultrasonication may be required.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Intended for research use only, not for clinical applications.

Catalog No. 50-002-39238 Supplier Medchemexpress LLC Supplier No. HY12481A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SAR405 R enantiomer is the less active enantiomer of SAR405, a selective inhibitor of PIK3C3/Vps34, supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO or as a solid, and is used as a reference or control in studies of autophagy, vesicle trafficking, and related signaling pathways.

  • Less active enantiomer of a PIK3C3/Vps34 inhibitor.
  • High purity (~99.9%) for reliable experimental controls.
  • Available as 10 mM solution in DMSO and multiple solid package sizes.
  • Soluble in DMSO at 100 mg/mL; ultrasonication may be required.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Intended for research use only, not for clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.