Learn More
Medchemexpress LLC SAR405 R enantiomer | 1946010-79-2 | 99.9% | 443.85 | C19H21ClF3N5O2 | 5MG
SDP
Catalog No. 5000239244
Encompass_Preferred
SAR405 R enantiomer is the less-active enantiomer of SAR405, a selective ATP-competitive inhibitor of class III PI3K (Vps34) used in research to profile autophagy and endosomal trafficking pathways. The compound is supplied as a high-purity solid and as a 10 mM solution in DMSO, with small pack sizes suitable for in vitro assays. Chemical identifiers: CAS 1946010-79-2; formula C19H21ClF3N5O2; molecular weight 443.85; purity 99.94%.
  • Less-active enantiomer for control and stereochemistry studies.
  • Selective inhibitor of PIK3C3/Vps34 for autophagy research.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as a 10 mM DMSO solution ready for use.
  • Small pack sizes enable screening and dose-response experiments.
  • Includes CAS and molecular data for compound tracking.
Catalog No. 50-002-39244 Supplier Medchemexpress LLC Supplier No. HY12481A5MG
SAR405 R enantiomer is the less-active enantiomer of SAR405, a selective ATP-competitive inhibitor of class III PI3K (Vps34) used in research to profile autophagy and endosomal trafficking pathways. The compound is supplied as a high-purity solid and as a 10 mM solution in DMSO, with small pack sizes suitable for in vitro assays. Chemical identifiers: CAS 1946010-79-2; formula C19H21ClF3N5O2; molecular weight 443.85; purity 99.94%.
  • Less-active enantiomer for control and stereochemistry studies.
  • Selective inhibitor of PIK3C3/Vps34 for autophagy research.
  • High purity suitable for biochemical and cell-based assays.
  • Available as solid and as a 10 mM DMSO solution ready for use.
  • Small pack sizes enable screening and dose-response experiments.
  • Includes CAS and molecular data for compound tracking.