Learn More
TARGETMOL CHEMICALS INC SAR405 R enantiomer 5MG
SDP
Catalog No. 502875911
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.SAR405 (R enantiomer) is the less active form of SAR405, an inhibitor of PIK3C3/Vps34. Purity 98.04%
Catalog No. 50-287-5911 Supplier TARGETMOL CHEMICALS INC Supplier No. T128315MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.SAR405 (R enantiomer) is the less active form of SAR405, an inhibitor of PIK3C3/Vps34. Purity 98.04%