Learn More
Selleck Chemical LLC SAR405838
SDP
Catalog No. 5000004839
Encompass_Preferred
SAR405838 is an orally available MDM2 antagonist with Ki of 0 88 nM SAR405838 (CAS 1303607-60-4) is a diastereomer of MI-773 (CAS 1303607-07-9) Phase 1
Catalog No. 50-000-04839 Supplier Selleck Chemical LLC Supplier No. S764925MG
SAR405838 is an orally available MDM2 antagonist with Ki of 0 88 nM SAR405838 (CAS 1303607-60-4) is a diastereomer of MI-773 (CAS 1303607-07-9) Phase 1