Learn More
TARGETMOL CHEMICALS INC SAR407899 25MG
SDP
Catalog No. 502579198
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAR407899 is Rho kinase inhibitor potently inhibits endothelin-1-induced constriction of renal resistance arteries. purity: 99%
Catalog No. 50-257-9198 Supplier TARGETMOL CHEMICALS INC Supplier No. T739125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAR407899 is Rho kinase inhibitor potently inhibits endothelin-1-induced constriction of renal resistance arteries. purity: 99%