Learn More
Medchemexpress LLC SAR407899 (hydrochloride) | 923262-96-8 | 99.1% | 280.75 | 5 MG
SDP
Catalog No. 5000364195
Encompass_Preferred
SAR407899 hydrochloride is a selective, potent, and ATP-competitive ROCK inhibitor. It has an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively. It inhibits ROCK-2 more effectively than ROCK-1. This product exhibits potent vasorelaxant activity in various isolated arteries and dose-dependently lowers blood pressure in hypertensive SHRs. It also increases penile length in healthy and diabetic rabbits.
  • Selective and potent ROCK inhibitor
  • ATP-competitive mechanism of action
  • Inhibits MYPTT696 phosphorylation
  • Blocks thrombin-induced effects
  • Inhibits cell proliferation and migration
  • Potent vasorelaxant activity
  • Reduces blood pressure
  • Increases penile length
Catalog No. 50-003-64195 Supplier Medchemexpress LLC Supplier No. 5MGHY15687
SAR407899 hydrochloride is a selective, potent, and ATP-competitive ROCK inhibitor. It has an IC50 of 135 nM for ROCK-2, and Kis of 36 nM and 41 nM for human and rat ROCK-2, respectively. It inhibits ROCK-2 more effectively than ROCK-1. This product exhibits potent vasorelaxant activity in various isolated arteries and dose-dependently lowers blood pressure in hypertensive SHRs. It also increases penile length in healthy and diabetic rabbits.
  • Selective and potent ROCK inhibitor
  • ATP-competitive mechanism of action
  • Inhibits MYPTT696 phosphorylation
  • Blocks thrombin-induced effects
  • Inhibits cell proliferation and migration
  • Potent vasorelaxant activity
  • Reduces blood pressure
  • Increases penile length