Learn More
Medchemexpress LLC Sar441255 TFA | 99.9% | 4866.56 (free base) | 1 MG
SDP
Catalog No. 5000424404
Encompass_Preferred
SAR441255 TFA is a potent unimolecular peptide GLP-1/GIP/GCG receptor triagonist. It demonstrates high potency with balanced activation of all three target receptors. This compound shows positive acute glucoregulatory effects in diabetic obese monkeys, indicating its potential for metabolic and endocrine disease research, particularly in areas like obesity and type 2 diabetes.
  • Potent unimolecular peptide receptor triagonist.
  • Displays high potency with balanced activation of three target receptors.
  • Shows positive acute glucoregulatory effects in diabetic obese monkeys.
  • Relevant for research in metabolic diseases, including obesity and type 2 diabetes.
Catalog No. 50-004-24404 Supplier Medchemexpress LLC Supplier No. HYP10031A1MG
SAR441255 TFA is a potent unimolecular peptide GLP-1/GIP/GCG receptor triagonist. It demonstrates high potency with balanced activation of all three target receptors. This compound shows positive acute glucoregulatory effects in diabetic obese monkeys, indicating its potential for metabolic and endocrine disease research, particularly in areas like obesity and type 2 diabetes.
  • Potent unimolecular peptide receptor triagonist.
  • Displays high potency with balanced activation of three target receptors.
  • Shows positive acute glucoregulatory effects in diabetic obese monkeys.
  • Relevant for research in metabolic diseases, including obesity and type 2 diabetes.