Learn More
Cayman Chemical SAR441255sodIum salt 10mg
SDP
Catalog No. 502944810
Encompass_Preferred
A peptide agonist of GLP-1R, GIPR, and GCGR; induces cAMP accumulation in HEK293 cells expressing human GLP-1R, GIPR, or GCGR (EC50s = 1.03, 0.73, and 1.01 pM, respectively); reduces body weight in a mouse model of high-fat diet-induced obesity from 1-30 UG/kg; reduces body weight and blood HbA1c levels in high-fat diet-induced obese diabetic cynomolgus monkeys at 11 UG/kg
Catalog No. 50-294-4810 Supplier Cayman Chemical Supplier No. 4117510 MG
A peptide agonist of GLP-1R, GIPR, and GCGR; induces cAMP accumulation in HEK293 cells expressing human GLP-1R, GIPR, or GCGR (EC50s = 1.03, 0.73, and 1.01 pM, respectively); reduces body weight in a mouse model of high-fat diet-induced obesity from 1-30 UG/kg; reduces body weight and blood HbA1c levels in high-fat diet-induced obese diabetic cynomolgus monkeys at 11 UG/kg