Learn More
Medchemexpress LLC HY-137472 5mg Medchemexpress, SAR502250 CAS:503860-57-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374725MG
Encompass_Preferred
Medchemexpress, HY-137472 5mg SAR502250 CAS:503860-57-9 SAR502250 is a potent, selective, ATP competitive, orally active and brain-penetrant inhibitor of GSK3, with an IC50 of 12 nM for human GSK-3β. SAR502250 displays antidepressant-like activity. SAR502250 can be used for the research of Alzheimer’s disease (AD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1491