Learn More
Apexbio Technology LLC SAR7334 1333210-07-3 2mg
SDP
Catalog No. 502240371
Encompass_Preferred
SAR7334 (CAS 1333210-07-3) is a small molecule inhibitor that selectively targets the transient receptor potential cation channel subfamily C member 6 (TRPC6) By inhibiting TRPC6-mediated calcium influx SAR7334 is utilized to investigate TRPC6-related signal transduction pathways in cellular and preclinical studies This compound serves as a valuable tool for elucidating the role of TRPC6 in pathophysiological processes including those related to kidney function and vascular regulation Its application aids in advancing the understanding of TRPC6-associated diseases and potential therapeutic interventions
This item is not returnable. View return policy
Catalog No. 50-224-0371 Supplier Apexbio Technology LLC Supplier No. B11892
SAR7334 (CAS 1333210-07-3) is a small molecule inhibitor that selectively targets the transient receptor potential cation channel subfamily C member 6 (TRPC6) By inhibiting TRPC6-mediated calcium influx SAR7334 is utilized to investigate TRPC6-related signal transduction pathways in cellular and preclinical studies This compound serves as a valuable tool for elucidating the role of TRPC6 in pathophysiological processes including those related to kidney function and vascular regulation Its application aids in advancing the understanding of TRPC6-associated diseases and potential therapeutic interventions