Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SAR7334 hydrochloride | 1333207-63-8 | 98.0% | 440.79 g/mol | C21H24Cl3N3O | 10 MG
SDP

Catalog No. 5000287874
Encompass_Preferred

SAR7334 hydrochloride is the hydrochloride salt of SAR7334, a potent and selective small-molecule inhibitor of the TRPC6 ion channel. It inhibits TRPC6 currents with an IC50 of 7.9 nM and is supplied as a high-purity research reagent for biochemical and cellular studies.

  • Potent TRPC6 inhibitor (IC50 7.9 nM).
  • Hydrochloride salt form for improved stability and solubility.
  • Purity: 98.0% (as supplied).
  • Molecular weight: 440.79 g/mol.
  • Available as milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-87874 Supplier Medchemexpress LLC Supplier No. HY15699A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SAR7334 hydrochloride is the hydrochloride salt of SAR7334, a potent and selective small-molecule inhibitor of the TRPC6 ion channel. It inhibits TRPC6 currents with an IC50 of 7.9 nM and is supplied as a high-purity research reagent for biochemical and cellular studies.

  • Potent TRPC6 inhibitor (IC50 7.9 nM).
  • Hydrochloride salt form for improved stability and solubility.
  • Purity: 98.0% (as supplied).
  • Molecular weight: 440.79 g/mol.
  • Available as milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.