Learn More
TARGETMOL CHEMICALS INC SAR7334 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581414
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAR7334 hydrochloride is a potent and specific inhibitor of TRPC6(IC50 of 7.9 nM). purity: 98%
Catalog No. 50-258-1414 Supplier TARGETMOL CHEMICALS INC Supplier No. T12849L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SAR7334 hydrochloride is a potent and specific inhibitor of TRPC6(IC50 of 7.9 nM). purity: 98%