Learn More
Medchemexpress LLC HY-P1012 1mg Medchemexpress, [Sar9,Met(O2)11]-Substance P CAS:110880-55-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10121MG
Encompass_Preferred
Medchemexpress, HY-P1012 1mg [Sar9,Met(O2)11]-Substance P CAS:110880-55-2 [Sar9,Met(O2)11]-Substance P is a tachykinin NK 1 receptor selective agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4127