Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [Sar9,Met(O2)11]-Substance P | 110880-55-2 | 99.6% | 1393.66 | 10 MG
SDP

Catalog No. 5000423711
Encompass_Preferred

[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.

  • For research use only
  • Offered in various quantities
  • Available in solid form, white to off-white color
  • Can be shipped at room temperature within the continental US
  • Recommended storage: sealed storage, away from moisture; powder at -80°C for 2 years or -20°C for 1 year. In solvent: -80°C for 6 months or -20°C for 1 month
  • Solubility in H2O: 100 mg/mL (71.75 mM)

Catalog No. 50-004-23711 Supplier Medchemexpress LLC Supplier No. HYP101210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.

  • For research use only
  • Offered in various quantities
  • Available in solid form, white to off-white color
  • Can be shipped at room temperature within the continental US
  • Recommended storage: sealed storage, away from moisture; powder at -80°C for 2 years or -20°C for 1 year. In solvent: -80°C for 6 months or -20°C for 1 month
  • Solubility in H2O: 100 mg/mL (71.75 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.