Learn More
Medchemexpress LLC [Sar9,Met(O2)11]-Substance P | 110880-55-2 | 99.6% | 1393.66 | 25 MG
SDP
Catalog No. 5000426299
Encompass_Preferred
[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.
  • Target: NK1 receptor
  • Cell line: U-373MG ATCC
  • IC50 value: 1.1 x 10-10 M
  • Increases mean arterial blood pressure and heart rate
  • Increases face washing and sniffing
  • Produces grooming
Catalog No. 50-004-26299 Supplier Medchemexpress LLC Supplier No. 25MGHYP1012
[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.
  • Target: NK1 receptor
  • Cell line: U-373MG ATCC
  • IC50 value: 1.1 x 10-10 M
  • Increases mean arterial blood pressure and heart rate
  • Increases face washing and sniffing
  • Produces grooming