Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [Sar9,Met(O2)11]-Substance P | 110880-55-2 | 99.6% | 1393.66 | 5 MG
SDP

Catalog No. 5000424719
Encompass_Preferred

[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.

  • High purity: 99.6%
  • Appearance: White to off-white solid
  • Molecular weight: 1393.66
  • Soluble in H2O and PBS at 100 mg/mL with sonication
  • Powder stability: 2 years at -80°C, 1 year at -20°C (sealed, away from moisture)
  • In-solvent stability: 6 months at -80°C, 1 month at -20°C (sealed, away from moisture)
  • Targets NK1 receptor with an IC50 of 1.1 x 10-10 M in human U373MG cells

Catalog No. 50-004-24719 Supplier Medchemexpress LLC Supplier No. HYP10125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Sar9,Met(O2)11]-Substance P is a tachykinin NK1 receptor selective agonist.

  • High purity: 99.6%
  • Appearance: White to off-white solid
  • Molecular weight: 1393.66
  • Soluble in H2O and PBS at 100 mg/mL with sonication
  • Powder stability: 2 years at -80°C, 1 year at -20°C (sealed, away from moisture)
  • In-solvent stability: 6 months at -80°C, 1 month at -20°C (sealed, away from moisture)
  • Targets NK1 receptor with an IC50 of 1.1 x 10-10 M in human U373MG cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.