Learn More
Medchemexpress LLC [Sar9] Substance P | 77128-75-7 | >98% | 1 MG
SDP
Catalog No. 5000424449
Encompass_Preferred
[Sar9] Substance P is a potent and selective neurokinin (NK)-1 receptor agonist. This synthetic neuropeptide analogue activates the NK-1 receptor, making it a valuable tool for studying neurokinin receptor-mediated pathways.
  • Potent NK-1 receptor agonist
  • Selective for NK-1 receptor
  • Useful for neurokinin receptor research
Catalog No. 50-004-24449 Supplier Medchemexpress LLC Supplier No. HYP17381MG
[Sar9] Substance P is a potent and selective neurokinin (NK)-1 receptor agonist. This synthetic neuropeptide analogue activates the NK-1 receptor, making it a valuable tool for studying neurokinin receptor-mediated pathways.
  • Potent NK-1 receptor agonist
  • Selective for NK-1 receptor
  • Useful for neurokinin receptor research