Learn More
Cayman Chemical SaracatInIb 10mg
SDP
Catalog No. 502951694
Encompass_Preferred
A dual inhibitor of c-Src and Abl (IC50 = 2.7 and 30 nM, respectively); less effectively inhibits other kinases; blocks cell motility, migration, adhesion, invasion, proliferation, differentiation, and survival; reduces osteoclast bone resorption
Catalog No. 50-295-1694 Supplier Cayman Chemical Supplier No. 1149710 MG
A dual inhibitor of c-Src and Abl (IC50 = 2.7 and 30 nM, respectively); less effectively inhibits other kinases; blocks cell motility, migration, adhesion, invasion, proliferation, differentiation, and survival; reduces osteoclast bone resorption