Learn More
Medchemexpress LLC HY-10234 50mg , Saracatinib CAS:379231-04-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023450MG
Encompass_Preferred
HY-10234 50mg Saracatinib CAS: 379231-04-6 Saracatinib (AZD0530) is a potent Src family inhibitor with IC50s of 2.7 to 11 nM for c-Src, Lck, c-YES, Lyn, Fyn, Fgr, and Blk. Saracatinib shows high selectivity over other tyrosine kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9897