Learn More
Cayman Chemical Saracatinib, Purity: ≥90%, CAS: 379231-04-6, 50 mg
SDP
Catalog No. 502952108
Encompass_Preferred
A dual inhibitor of c-Src and Abl (IC50 = 2.7 and 30 nM, respectively); less effectively inhibits other kinases; blocks cell motility, migration, adhesion, invasion, proliferation, differentiation, and survival; reduces osteoclast bone resorption
Catalog No. 50-295-2108 Supplier Cayman Chemical Supplier No. 1149750 MG
A dual inhibitor of c-Src and Abl (IC50 = 2.7 and 30 nM, respectively); less effectively inhibits other kinases; blocks cell motility, migration, adhesion, invasion, proliferation, differentiation, and survival; reduces osteoclast bone resorption