Learn More
Selleck Chemical LLC Saracatinib (AZD0530) 200mg 379231-04-6
SDP
Catalog No. 507781
Encompass_Preferred
Saracatinib (AZD0530) is a potent Src inhibitor with IC50 of 2.7 nM in cell-free assays, and potent to c-Yes, Fyn, Lyn, Blk, Fgr and Lck; less active for Abl and EGFR (L858R and L861Q). Saracatinib induces autophagy. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-778-1 Supplier Selleck Chemical LLC Supplier No. S1006200MG
Saracatinib (AZD0530) is a potent Src inhibitor with IC50 of 2.7 nM in cell-free assays, and potent to c-Yes, Fyn, Lyn, Blk, Fgr and Lck; less active for Abl and EGFR (L858R and L861Q). Saracatinib induces autophagy. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.