Learn More
Cayman Chemical Sarafloxacin-d8 hydrochlorid
SDP
Catalog No. 502949350
Encompass_Preferred
An internal standard for the quantification of sarafloxacin by GC- or LC-MS
Catalog No. 50-294-9350 Supplier Cayman Chemical Supplier No. 34226500 UG
An internal standard for the quantification of sarafloxacin by GC- or LC-MS