Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarafloxacin HCl Trihydrate, G-Biosciences
SCI_ED

Catalog No. S000019
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000019 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000019 Supplier G-Biosciences Supplier No. RC1098
Add to Cart
Add to Cart

Sarafloxacin is a broad-spectrum fluoroquinolone antibiotic effective against a variety of gram-negative and gram-positive organisms, especially Mycobacterium tuberculosis.

Sarafloxacin HCl Trihydrate is water soluble.

Chemical Identifiers

CAS 91296-87-6
Molecular Weight (g/mol) 475.87 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 25 g
Percent Purity 98%
Grade Molecular Biology
Chemical Name or Material Sarafloxacin HCl Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.