Learn More
Medchemexpress LLC Sarafloxacin (hydrochloride) | 91296-87-6 | 1 ML
SDP
Catalog No. 5000406210
Encompass_Preferred
Sarafloxacin hydrochloride is a quinolone antibiotic agent primarily used in veterinary medicine. It functions by inhibiting bacterial DNA gyrase, effectively suppressing bacterial growth. This antibiotic is also utilized in animal feed to enhance feed conversion and support growth.
  • Quinolone antibiotic agent.
  • Inhibits bacterial DNA gyrase.
  • Used in veterinary medicine to inhibit bacterial infections.
  • Improves feed conversion and promotes growth in animal feed.
  • Registered for use against poultry diseases.
  • Efficacious in treating channel catfish infected with *E. ictaluri*.
Catalog No. 50-004-06210 Supplier Medchemexpress LLC Supplier No. HYB0343A10MM/1ML
Sarafloxacin hydrochloride is a quinolone antibiotic agent primarily used in veterinary medicine. It functions by inhibiting bacterial DNA gyrase, effectively suppressing bacterial growth. This antibiotic is also utilized in animal feed to enhance feed conversion and support growth.
  • Quinolone antibiotic agent.
  • Inhibits bacterial DNA gyrase.
  • Used in veterinary medicine to inhibit bacterial infections.
  • Improves feed conversion and promotes growth in animal feed.
  • Registered for use against poultry diseases.
  • Efficacious in treating channel catfish infected with *E. ictaluri*.