Learn More
Medchemexpress LLC Sarafloxacin hydrochloride | 91296-87-6 | 98.13% | 100 MG
SDP
Catalog No. 5000406209
Encompass_Preferred
Sarafloxacin hydrochloride is a quinolone antibiotic agent registered for use against poultry diseases. It has been shown to be effective in treating channel catfish infected with *E. ictaluri*.
  • Quinolone antibiotic agent
  • Registered for use against poultry diseases
  • Efficacious in treating channel catfish infected with *E. ictaluri*
  • Inhibitory level is strain dependent for tested bacteria
Catalog No. 50-004-06209 Supplier Medchemexpress LLC Supplier No. HYB0343A100MG
Sarafloxacin hydrochloride is a quinolone antibiotic agent registered for use against poultry diseases. It has been shown to be effective in treating channel catfish infected with *E. ictaluri*.
  • Quinolone antibiotic agent
  • Registered for use against poultry diseases
  • Efficacious in treating channel catfish infected with *E. ictaluri*
  • Inhibitory level is strain dependent for tested bacteria